missing translation for 'onlineSavingsMsg'
Learn More
4-Azabenzimidazole, 98%, Thermo Scientific Chemicals Product Code.: 11436027

4-Azabenzimidazole, 98%, Thermo Scientific Chemicals

Product Code. 11436027
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11436027

Brand: Thermo Scientific Alfa Aesar B25051.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-Azabenzimidazole is used in the preparation of imidazo[4,5-b]pyridine 4-oxide. It is also used as a reagent to prepare benzoylmethyl-substituted azoles which are inhibitors of nitric oxide synthase with antioxidant properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Azabenzimidazole is used in the preparation of imidazo[4,5-b]pyridine 4-oxide. It is also used as a reagent to prepare benzoylmethyl-substituted azoles which are inhibitors of nitric oxide synthase with antioxidant properties.

Solubility
Soluble in dimethylformamide.

Notes
Incompatible with strong oxidizing agent and strong acid.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 273-21-2
Molecular Formula C6H5N3
Molecular Weight (g/mol) 119.13
MDL Number MFCD00005579,MFCD01646138
InChI Key GAMYYCRTACQSBR-UHFFFAOYSA-N
Synonym 3h-imidazo 4,5-b pyridine, 4-azabenzimidazole, 1h-imidazo 4,5-b pyridine, 1-deazapurine, 3,4-diazaindole, 7-azabenzimidazole, imidazopyridine, 4h-imidazo 4,5-b pyridine, pyrido 2,3-d imidazole, azabenzimidazole
PubChem CID 67504
ChEBI CHEBI:59952
IUPAC Name 1H-imidazo[4,5-b]pyridine
SMILES N1C=NC2=NC=CC=C12

Specifications

Melting Point 145°C to 150°C
Boiling Point 140°C to 150°C (0.1 mmHg)
Quantity 5 g
Beilstein 2600
Solubility Information Soluble in dimethylformamide.
Formula Weight 119.13
Percent Purity 99%
Chemical Name or Material 4-Azabenzimidazole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberJ7928A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25051.06,B25051.03,B25051.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Azabenzimidazole, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.