missing translation for 'onlineSavingsMsg'
Learn More

4-Benzoylbutyric acid, 97%, Thermo Scientific Chemicals

Product Code. 10773963
Click to view available options
25g; Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1501-05-9
CAS Min % 96.0
CAS Max % 100.0
Melting Point 124°C to 128°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C11H12O3
Linear Formula C6H5CO(CH2)3CO2H
MDL Number MFCD00004411
Quantity 25 g
Beilstein 10, 708
Synonym 4-benzoylbutyric acid, 5-oxo-5-phenylvaleric acid, benzenepentanoic acid, .delta.-oxo, 4-benzoyl-butyric acid, timtec-bb sbb007811, 5-oxo-5-phenyl-pentanoic acid, 4-benzoylbutyric acid;4-benzoylbutanoic acid, butanoic acid, 4-benzoyl, 4-benzoylbutyricacid
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in benzene (warm),chloroform,ethanol,,dichloromethane,ethyl acetate,aqueous alkali,insoluble in hexane
InChI Key SHKWSBAVRQZYLE-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C(=O)CCCC(=O)O
IUPAC Name 5-oxo-5-phenylpentanoic acid
Molecular Weight (g/mol) 192.21
PubChem CID 73914
Formula Weight 192.21
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzoylbutyric acid, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Benzoylbutyric acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.