missing translation for 'onlineSavingsMsg'
Learn More

4-Benzoylpiperidine hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 10296670
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25519-80-6
Molecular Formula C12H15NO·ClH
Molecular Weight (g/mol) 225.72
MDL Number MFCD00066982
InChI Key NXYKIFZJQXOUJS-UHFFFAOYSA-N
Synonym 4-benzoylpiperidine hydrochloride, phenyl piperidin-4-yl methanone hydrochloride, 4-benzoylpiperidine hcl, phenyl 4-piperidyl methanone hydrochloride, 4-benzoyl piperidine hydrochloride, 4-benzoylpiperidine hydrogen chloride, methanone, phenyl-4-piperidinyl-, hydrochloride, phenyl 4-piperidyl ketone, chloride, 4-benzoyl piperidine hcl, pubchem9881
PubChem CID 2724437
IUPAC Name phenyl(piperidin-4-yl)methanone;hydrochloride
SMILES C1CNCCC1C(=O)C2=CC=CC=C2.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 226°C to 227°C
Assay Percent Range 98%
Packaging Glass bottle
Quantity 10 g
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in methanol and ethanol
Formula Weight 225.72
Percent Purity 98%
Chemical Name or Material 4-Benzoylpiperidine hydrochloride, 98%
compliance-icons
Product Identifier
  • 4-Benzoylpiperidine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'