missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-3-chlorobenzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15439807
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 845551-44-2
Molecular Formula C13H12BClO3
Molecular Weight (g/mol) 262.496
MDL Number MFCD04115642
InChI Key AYSMMNDQVZAXQY-UHFFFAOYSA-N
Synonym 4-benzyloxy-3-chlorophenylboronic acid, 4-benzyloxy-3-chlorophenyl boronic acid, 3-chloro-4-benzyloxyphenyl boronic acid, 4-benzyloxy-3-chlorobenzeneboronic acid, boronic acid, 3-chloro-4-phenylmethoxy phenyl, b-3-chloro-4-phenylmethoxy phenyl-boronic acid, akos brn-0594, acmc-209pvk, 4-benzyloxy-3-chlorophenylboro
PubChem CID 3763657
IUPAC Name (3-chloro-4-phenylmethoxyphenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)OCC2=CC=CC=C2)Cl)(O)O

Specifications

Melting Point 154°C to 162°C
Quantity 5 g
Formula Weight 262.5
Percent Purity 98%
Chemical Name or Material 4-Benzyloxy-3-chlorobenzeneboronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.