missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-3-fluorobenzeneboronic acid, 98%, Thermo Scientific™

Product Code. 11315035
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133057-83-7
Molecular Formula C13H12BFO3
Molecular Weight (g/mol) 246.044
MDL Number MFCD00994627
InChI Key DQPDLDQJMUGVGI-UHFFFAOYSA-N
Synonym 4-benzyloxy-3-fluorophenylboronic acid, 3-fluoro-4-benzyloxyphenyl boronic acid, 4-benzyloxy-3-fluorobenzeneboronic acid, 4-benzyloxy-3-fluorophenyl boronic acid, 4-benzyloxy-3-fluorophenyl boranediol, boronic acid, 3-fluoro-4-phenylmethoxy phenyl, pubchem5322
PubChem CID 2737788
IUPAC Name (3-fluoro-4-phenylmethoxyphenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)OCC2=CC=CC=C2)F)(O)O

Specifications

Melting Point 129°C to 131°C
Quantity 1 g
Formula Weight 246.04
Percent Purity 98%
Chemical Name or Material 4-Benzyloxy-3-fluorobenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'