missing translation for 'onlineSavingsMsg'
Learn More
4-(Benzyloxycarbonylamino)cyclohexanone, 97%, Thermo Scientific™ Product Code.: 11341929

4-(Benzyloxycarbonylamino)cyclohexanone, 97%, Thermo Scientific™

Product Code. 11341929
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11341929

Brand: Thermo Scientific Alfa Aesar H26015.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16801-63-1
Molecular Formula C14H17NO3
Molecular Weight (g/mol) 247.29
MDL Number MFCD06658393
InChI Key VHXBIBFCJISKFA-UHFFFAOYSA-N
Synonym benzyl 4-oxocyclohexyl carbamate, 4-n-cbz-cyclohexanone, 4-n-cbz-amino-cyclohexanone, 4-oxocyclohexyl carbamic acid benzyl ester, 4-n-cbz-aminocyclohexanone, 4-benzyloxycarbonylamino cyclohexanone, 4-cbz-amino cyclohexanone, n-cbz-4-amino-cyclohexanone, benzyl n-4-oxocyclohexyl carbamate, n-cbz-4-aminocyclohexanone
PubChem CID 14615292
IUPAC Name benzyl N-(4-oxocyclohexyl)carbamate
SMILES O=C(NC1CCC(=O)CC1)OCC1=CC=CC=C1

Specifications

Melting Point 81°C to 83°C
Odor Odorless
Quantity 1 g
Formula Weight 247.29
Percent Purity 97%
Chemical Name or Material 4-(Benzyloxycarbonylamino)cyclohexanone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(Benzyloxycarbonylamino)cyclohexanone, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.