missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxyphenoxyacetic acid, 95%, Thermo Scientific™

Product Code. 11374424
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38559-92-1
Molecular Formula C15H14O4
Molecular Weight (g/mol) 258.273
MDL Number MFCD00014361
InChI Key VXMSXBVTUNOSLL-UHFFFAOYSA-N
Synonym 4-benzyloxyphenoxyacetic acid, 4-benzyloxy phenoxy acetic acid, 4-benzyloxy phenoxyacetic acid, 2-4-benzyloxy phenoxy acetic acid, 4-phenylmethoxy phenoxy acetic acid, acetic acid, 4-phenylmethoxy phenoxy, 2-4-phenylmethoxy phenoxy acetic acid, enamine_002147, acmc-1agp8
PubChem CID 563696
IUPAC Name 2-(4-phenylmethoxyphenoxy)acetic acid
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)OCC(=O)O

Specifications

Melting Point 139°C to 140°C
Quantity 50 g
Beilstein 2469260
Formula Weight 258.28
Percent Purity 95%
Chemical Name or Material 4-Benzyloxyphenoxyacetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'