missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxyphenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 10713445
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52068-30-1
Molecular Formula C13H14N2O·HCl
Molecular Weight (g/mol) 250.73
MDL Number MFCD00137741
InChI Key OVNUPJXMCMTQCN-UHFFFAOYSA-N
Synonym 4-benzyloxyphenylhydrazine hydrochloride, 4-benzyloxy phenyl hydrazine hydrochloride, 4-benzyloxyphenyl hydrazine hydrochloride, 4-benzyloxy phenyl hydrazine monohydrochloride, 4-benzyloxyphenyl hydrazine hcl, 4-benzyloxyphenylhydrazine hcl, 4-benzyloxyphenylhydrazinehydrochloride, 4-phenylmethoxyphenyl hydrazine hydrochloride
PubChem CID 3016725
IUPAC Name (4-phenylmethoxyphenyl)hydrazine;hydrochloride
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)NN.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 187°C to 189°C
Color Brown to Cream
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH2OC6H4NHNH2·HCl
Quantity 1 g
Formula Weight 250.73
Percent Purity 98%
Physical Form Fluffy Powder
Chemical Name or Material 4-Benzyloxyphenylhydrazine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 4-Benzyloxyphenylhydrazine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'