missing translation for 'onlineSavingsMsg'
Learn More

4-Benzylpiperazine-1-carboximidamide hydroiodide, 95%, Thermo Scientific™

Product Code. 10387644
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 852228-15-0
Molecular Formula C12H19IN4
Molecular Weight (g/mol) 346.22
MDL Number MFCD09025921
InChI Key CXQITJKGJPFOIV-UHFFFAOYSA-N
Synonym 4-benzylpiperazine-1-carboximidamide hydroiodide, 4-benzylpiperazine-1-carboximidamide-hydrogen iodide 1/1
PubChem CID 18525929
IUPAC Name 4-benzylpiperazine-1-carboximidamide;hydroiodide
SMILES I.NC(=N)N1CCN(CC2=CC=CC=C2)CC1

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 346.21
Percent Purity 95%
Chemical Name or Material 4-Benzylpiperazine-1-carboximidamide hydroiodide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'