missing translation for 'onlineSavingsMsg'
Learn More

4-(Boc-amino)benzoic acid, 97%, Thermo Scientific Chemicals

Product Code. 11399385
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66493-39-8
Molecular Formula C12H15NO4
Molecular Weight (g/mol) 237.255
MDL Number MFCD00037428
InChI Key ZJDBQMWMDZEONW-UHFFFAOYSA-N
Synonym boc-4-abz-oh, 4-boc-amino benzoic acid, 4-tert-butoxycarbonyl amino benzoic acid, 4-tert-butoxycarbonylamino-benzoic acid, n-boc-4-aminobenzoic acid, 4-tert-butoxycarbonylaminobenzoic acid, 4-tert-butoxycarbonylamino benzoic acid, boc-p-aminobenzoic acid, 4-boc-amino-benzoic acid
PubChem CID 2755931
IUPAC Name 4-[(2-methylpropan-2-yl)oxycarbonylamino]benzoic acid
SMILES CC(C)(C)OC(=O)NC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point ∽193°C (decomposition)
Quantity 1 g
Beilstein 2115614
Formula Weight 237.26
Percent Purity 97%
Chemical Name or Material 4-(Boc-amino)benzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.