missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-1,3-benzodioxole, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 4-bromobenzo d 1,3 dioxole, 4-bromo-2h-1,3-benzodioxole, 4-bromo-benzo 1,3 dioxole, 1,3-benzodioxole, 4-bromo, 2,3-methylenedioxy-bromobenzene, 4-bromo-1,3-benzodioxol, methylene dioxy bromo-benzene, 1,3-benzodioxole,4-bromo, 4-bromanyl-1,3-benzodioxole, 1,3-benzodioxol-4-yl bromide |
Formula Weight | 201.02 |
Percent Purity | 97% |
Chemical Name or Material | 4-Bromo-1, 3-benzodioxole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'