missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-butene, 98+%, Thermo Scientific Chemicals

70.45 € - 395.97 €

Chemical Identifiers

CAS 5162-44-7
Molecular Formula C4H7Br
Molecular Weight (g/mol) 135
MDL Number MFCD00000258
InChI Key DMAYBPBPEUFIHJ-UHFFFAOYSA-N
Synonym 4-bromo-1-butene, 3-butenyl bromide, 1-butene, 4-bromo, 4-bromo-1-buten, 4-bromo-but-1-ene, homoallyl bromide, 1-bromo-3-butene, 4-bromobutene, 4-bromo-1 butene, sjphabijuh@
PubChem CID 21241
IUPAC Name 4-bromobut-1-ene
SMILES C=CCCBr
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10523351
View Documents
Thermo Scientific Acros
173550050
5 mL Glass bottle
70.45 €
5mL
Please to purchase this item. Need a web account? Register with us today!
10533351
View Documents
Thermo Scientific Acros
173550250
25 mL Glass bottle
240.20 €
25mL
Please to purchase this item. Need a web account? Register with us today!
10543351
View Documents
Thermo Scientific Acros
173550500
50 mL Glass bottle
395.97 €
50mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5162-44-7
135
DMAYBPBPEUFIHJ-UHFFFAOYSA-N
21241
C=CCCBr
C4H7Br
MFCD00000258
4-bromo-1-butene, 3-butenyl bromide, 1-butene, 4-bromo, 4-bromo-1-buten, 4-bromo-but-1-ene, homoallyl bromide, 1-bromo-3-butene, 4-bromobutene, 4-bromo-1 butene, sjphabijuh@
4-bromobut-1-ene

Specifications

5162-44-7
100.0
1.3300g/mL
1°C
98+%
BrCH2CH2CH=CH2
MFCD00000258
1.33
DMAYBPBPEUFIHJ-UHFFFAOYSA-N
4-bromobut-1-ene
21241
98+%
4-Bromo-1-butene, 98+%
98.0
Brown to Colorless
98.0°C to 100.0°C
Authentic
C4H7Br
1.4595 to 1.4635
01, I, 84
4-bromo-1-butene, 3-butenyl bromide, 1-butene, 4-bromo, 4-bromo-1-buten, 4-bromo-but-1-ene, homoallyl bromide, 1-bromo-3-butene, 4-bromobutene, 4-bromo-1 butene, sjphabijuh@
C=CCCBr
135
135
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
May cause an allergic skin reaction.
Highly flammable liquid and va

GHS P Statement
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear protective gloves/pr

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 225-937-4

missing translation for 'tsca' : TSCA

Product Identifier
  • 4-Bromo-1-butene

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.