missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-methyl-1H-imidazole, 95%, Thermo Scientific Chemicals

108.54 € - 320.60 €

Chemical Identifiers

CAS 25676-75-9
Molecular Formula C4H5BrN2
Molecular Weight (g/mol) 161.00
MDL Number MFCD01320501
InChI Key IOTSLMMLLXTNNH-UHFFFAOYSA-N
PubChem CID 1277653
IUPAC Name 4-bromo-1-methylimidazole
SMILES CN1C=NC(Br)=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15274914
View Documents
Thermo Scientific Acros
452430010
1 g
108.54 €
1g
Please to purchase this item. Need a web account? Register with us today!
15264914
View Documents
Thermo Scientific Acros
452430050
5 g
320.60 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

25676-75-9
161.00
IOTSLMMLLXTNNH-UHFFFAOYSA-N
4-bromo-1-methylimidazole
C4H5BrN2
MFCD01320501
1277653
CN1C=NC(Br)=C1

Specifications

25676-75-9
>110°C
Glass Bottle
MFCD01320501
IOTSLMMLLXTNNH-UHFFFAOYSA-N
4-bromo-1-methylimidazole
1277653
95%
91°C to 93°C (0.1 hPa)
94% min. (GC)
C4H5BrN2
4-bromo-1-methyl-1h-imidazole, 1h-imidazole, 4-bromo-1-methyl, acmc-1cjr4, 4-bromo-1-methyl-imidazole, 4-bromanyl-1-methyl-imidazole, 1-methyl-4-bromo-1h-imidazole, 1h-imidazole,4-bromo-1-methyl
CN1C=NC(Br)=C1
161.00
161
4-Bromo-1-methyl-1H-imidazole
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

Product Identifier
  • 4-Bromo-1-methyl-1H-imidazole, 95%

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.