missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-methyl-1H-pyrazole-5-carbaldehyde, ≥97%, Thermo Scientific™

46.70 € - 332.00 €

Chemical Identifiers

CAS 473528-88-0
Molecular Formula C5H5BrN2O
Molecular Weight (g/mol) 189.012
MDL Number MFCD03074341
InChI Key GXAHYXQWHWDEDY-UHFFFAOYSA-N
PubChem CID 2794634
IUPAC Name 4-bromo-2-methylpyrazole-3-carbaldehyde
SMILES CN1C(=C(C=N1)Br)C=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10039431
View Documents
Thermo Scientific Maybridge
MO00866CB
250 mg
46.70 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10212552
View Documents
Thermo Scientific Maybridge
MO00866DA
1 g
78.90 €
1g
Please to purchase this item. Need a web account? Register with us today!
10091784
View Documents
Thermo Scientific Maybridge
MO00866DE
5 g
332.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

473528-88-0
189.012
GXAHYXQWHWDEDY-UHFFFAOYSA-N
4-bromo-2-methylpyrazole-3-carbaldehyde
C5H5BrN2O
MFCD03074341
2794634
CN1C(=C(C=N1)Br)C=O

Specifications

473528-88-0
97%
C5H5BrN2O
4-bromo-1-methyl-1h-pyrazole-5-carbaldehyde, 4-bromo-1-methylpyrazole-5-carbaldehyde, 4-bromo-2-methyl-2h-pyrazole-3-carbaldehyde, 1h-pyrazole-5-carboxaldehyde, 4-bromo-1-methyl, acmc-1akfs, 4-bromo-1-methylpyrazole-5-carboxaldehyde, 4-bromo-1-methyl-1h-pyrazole-5-carboxaldehyde, 4-bromo-2-methyl-2 h-pyrazole-3-carbaldehyde
CN1C(=C(C=N1)Br)C=O
189.012
189.01
4-Bromo-1-methyl-1H-pyrazole-5-carbaldehyde
97.0
Amber glass bottle
MFCD03074341
GXAHYXQWHWDEDY-UHFFFAOYSA-N
4-bromo-2-methylpyrazole-3-carbaldehyde
2794634
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications