missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1H-pyrazole-5-carbaldehyde, 95+%, Thermo Scientific™

Product Code. 10550353
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 287917-97-9
Molecular Formula C4H3BrN2O
Molecular Weight (g/mol) 174.985
InChI Key UWGFONONBAIDAF-UHFFFAOYSA-N
Synonym 4-bromo-1h-pyrazole-3-carbaldehyde, 4-bromo-1h-pyrazole-5-carboxaldehyde, 4-bromo-2h-pyrazole-3-carbaldehyde, 4-bromo-1h-pyrazole-3-carboxaldehyde, 1h-pyrazole-3-carboxaldehyde,4-bromo, 1h-pyrazole-3-carboxaldehyde, 4-bromo, 4-bromopyrazole-5-carbaldehyde, 4-bromo-5-formyl-1h-pyrazole, 4-bromo-pyrazole-3-carboxaldehyde, 4-bromo-2h-pyrazole-3-carboxaldehyde
PubChem CID 2782154
IUPAC Name 4-bromo-1H-pyrazole-5-carbaldehyde
SMILES C1=NNC(=C1Br)C=O

Specifications

Assay Percent Range 95%
Quantity 250 mg
Formula Weight 174.98
Percent Purity 95%
Chemical Name or Material 4-Bromo-1H-pyrazole-5-carbaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.