missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-(1H-pyrazol-3-yl)phenol, 98%, Thermo Scientific™

Product Code. 11496620
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99067-15-9
Molecular Formula C9H7BrN2O
Molecular Weight (g/mol) 239.072
MDL Number MFCD03094026
InChI Key WAHULEHSSFNFQX-UHFFFAOYSA-N
Synonym 4-bromo-2-1h-pyrazol-3-yl phenol, 4-bromo-6-1,2-dihydropyrazol-3-ylidene cyclohexa-2,4-dien-1-one, 2-1h-pyrazole-3-yl-4-bromophenol, 4-bromo-2-2h-pyrazol-3-yl phenol
PubChem CID 6744501
IUPAC Name 4-bromo-6-(1,2-dihydropyrazol-3-ylidene)cyclohexa-2,4-dien-1-one
SMILES C1=CC(=O)C(=C2C=CNN2)C=C1Br

Spezifikation

Melting Point 159°C to 160°C
Quantity 25 g
Formula Weight 239.07
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-(1H-pyrazol-3-yl)phenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'