missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-cyanopyridine, 97%, Thermo Scientific Chemicals

Product Code. 15407377
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62150-45-2
Molecular Formula C6H3BrN2
Molecular Weight (g/mol) 183.008
MDL Number MFCD04065805
InChI Key CZXDCTUSFIKLIJ-UHFFFAOYSA-N
PubChem CID 693283
IUPAC Name 4-bromopyridine-2-carbonitrile
SMILES C1=CN=C(C=C1Br)C#N

Specifications

Melting Point 87°C to 91°C
Quantity 250 mg
UN Number UN3439
Synonym 4-bromo-2-cyanopyridine, 4-bromopicolinonitrile, 4-bromo-pyridine-2-carbonitrile, 2-pyridinecarbonitrile, 4-bromo, 2-cyano-4-bromopyridine, 4-bromo-2-pyridinecarbonitrile, pubchem23829, 4-bromo-2-cyano-pyridine, 2-pyridinecarbonitrile,4-bromo, abbypharma ap-17-5586
Formula Weight 183.01
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-cyanopyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'