missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromo-2'-hydroxy-5'-methylacetophenone, 97%, Thermo Scientific Chemicals

Product Code. 11452240
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1020253-81-9
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD08273792
InChI Key BPQABGQSVOUODJ-UHFFFAOYSA-N
Synonym 1-4-bromo-2-hydroxy-5-methylphenyl ethanone, 2-acetyl-4-methyl-5-bromophenol, 4'-bromo-2'-hydroxy-5'-methylacetophenone, 1-4-bromo-2-hydroxy-5-methylphenyl ethan-1-one, 1-acetyl-4-bromo-2-hydroxy-5-methylbenzene, 4'-bromo-2'-hydroxy-5'-methyl acetophenone
PubChem CID 42553093
IUPAC Name 1-(4-bromo-2-hydroxy-5-methylphenyl)ethanone
SMILES CC1=C(C=C(C(=C1)C(=O)C)O)Br

Specifications

Melting Point 86°C to 89°C
Quantity 1 g
Formula Weight 229.07
Percent Purity 97%
Chemical Name or Material 4'-Bromo-2'-hydroxy-5'-methylacetophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'