missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromo-2'-hydroxyacetophenone, 95%, Thermo Scientific Chemicals

Product Code. 15467638
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4'-Bromo-2'-hydroxyacetophenone is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Bromo-2′-hydroxyacetophenone is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30186-18-6
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD03428533
InChI Key LQCMMXGKEGWUIM-UHFFFAOYSA-N
Synonym 1-4-bromo-2-hydroxyphenyl ethanone, 4'-bromo-2'-hydroxyacetophenone, 4-bromo-2-hydroxyacetophenone, 1-4-bromo-2-hydroxyphenyl ethan-1-one, acetophenone,2-hydroxy-4-bromo, ethanone, 1-4-bromo-2-hydroxyphenyl, 2-acetyl-5-bromophenol, 1-4-bromo-2-hydroxy-phenyl ethanone, 1-4-bromo-2-hydroxy-phenyl-ethanone
PubChem CID 10727403
IUPAC Name 1-(4-bromo-2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=C(O)C=C(Br)C=C1

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 215.04
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4'-Bromo-2'-hydroxyacetophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'