missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Bromo-2'-hydroxyacetophenone, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
4'-Bromo-2'-hydroxyacetophenone is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4′-Bromo-2′-hydroxyacetophenone is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
4′-Bromo-2′-hydroxyacetophenone is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Specifications
Quantity | 250 mg |
Solubility Information | Slightly soluble in water. |
Formula Weight | 215.04 |
Percent Purity | 95% |
Physical Form | Solid |
Chemical Name or Material | 4'-Bromo-2'-hydroxyacetophenone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'