missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methoxyphenol, 98%, Thermo Scientific Chemicals

37.70 € - 460.00 €

Chemical Identifiers

CAS 7368-78-7
Molecular Formula C7H7BrO2
Molecular Weight (g/mol) 203.035
MDL Number MFCD00051937
InChI Key WHSIIJQOEGXWSN-UHFFFAOYSA-N
Synonym 4-bromoguaiacol, 5-bromo-2-hydroxyanisole, 4-bromo-2-methoxy-phenol, phenol, 4-bromo-2-methoxy, 2-methoxy-4-bromophenol, 5bromoguaiacol, 5-bromoguaiacol, 5-bromguajacol, pubchem17271, acmc-209orr
PubChem CID 262234
IUPAC Name 4-bromo-2-methoxyphenol
SMILES COC1=C(C=CC(=C1)Br)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11362716
View Documents
Thermo Scientific Alfa Aesar
L08801.04
2 g
37.70 €
2g
Please to purchase this item. Need a web account? Register with us today!
11372716
View Documents
Thermo Scientific Alfa Aesar
L08801.09
10 g
125.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
11382716
View Documents
Thermo Scientific Alfa Aesar
L08801.18
50 g
460.00 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

7368-78-7
203.035
WHSIIJQOEGXWSN-UHFFFAOYSA-N
262234
COC1=C(C=CC(=C1)Br)O
C7H7BrO2
MFCD00051937
4-bromoguaiacol, 5-bromo-2-hydroxyanisole, 4-bromo-2-methoxy-phenol, phenol, 4-bromo-2-methoxy, 2-methoxy-4-bromophenol, 5bromoguaiacol, 5-bromoguaiacol, 5-bromguajacol, pubchem17271, acmc-209orr
4-bromo-2-methoxyphenol

Specifications

7368-78-7
129°C to 132°C (12 mmHg)
C7H7BrO2
2045286
Slightly soluble in water.
COC1=C(C=CC(=C1)Br)O
203.035
203.04
4-Bromo-2-methoxyphenol
32°C to 37°C
Phenol-like
MFCD00051937
4-bromoguaiacol, 5-bromo-2-hydroxyanisole, 4-bromo-2-methoxy-phenol, phenol, 4-bromo-2-methoxy, 2-methoxy-4-bromophenol, 5bromoguaiacol, 5-bromoguaiacol, 5-bromguajacol, pubchem17271, acmc-209orr
WHSIIJQOEGXWSN-UHFFFAOYSA-N
4-bromo-2-methoxyphenol
262234
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.