missing translation for 'onlineSavingsMsg'
Learn More

(4-Bromo-2-thienyl)methanol, 97%, Thermo Scientific™

11.30 € - 27.40 €

Chemical Identifiers

CAS 79757-77-0
Molecular Formula C5H5BrOS
Molecular Weight (g/mol) 193.058
MDL Number MFCD04115392
InChI Key PXZNJHHUYJRFPZ-UHFFFAOYSA-N
PubChem CID 2795484
IUPAC Name (4-bromothiophen-2-yl)methanol
SMILES C1=C(SC=C1Br)CO
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10497232
View Documents
Thermo Scientific Maybridge
CC28509CB
250 mg Amber Glass Bottle
11.30 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10592642
View Documents
Thermo Scientific Maybridge
CC28509DA
1 g Amber Glass Bottle
27.40 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

79757-77-0
193.058
PXZNJHHUYJRFPZ-UHFFFAOYSA-N
(4-bromothiophen-2-yl)methanol
C5H5BrOS
MFCD04115392
2795484
C1=C(SC=C1Br)CO

Specifications

79757-77-0
100.0
C5H5BrOS
4-bromothiophen-2-yl methanol, 4-bromo-2-thienyl methanol, 4-bromo-2-hydroxymethylthiophene, 4-bromo-2-thiophenemethanol, 4-bromo-2-hydroxymethyl thiophene, 2-thiophenemethanol, 4-bromo, acmc-1bm1t, 3-bromo-5-thienyl methanol, 4-bromo-thiophene-2-methanol, 2-thiophenemethanol,4-bromo
C1=C(SC=C1Br)CO
193.058
193.06
(4-Bromo-2-thienyl)methanol
96.5
97%
MFCD04115392
PXZNJHHUYJRFPZ-UHFFFAOYSA-N
(4-bromothiophen-2-yl)methanol
2795484
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.