missing translation for 'onlineSavingsMsg'
Learn More
4-Bromo-2-(trifluoromethoxy)benzene-1-thiol, ≥95%, Thermo Scientific™ Product Code.: 10273282

4-Bromo-2-(trifluoromethoxy)benzene-1-thiol, ≥95%, Thermo Scientific™

Product Code. 10273282
10 g
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy

Product Code. 10273282

Brand: Thermo Scientific Maybridge TL00665EA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175278-15-6
Molecular Formula C7H4BrF3OS
Molecular Weight (g/mol) 273.07
MDL Number MFCD00203474
InChI Key WWZNHDMEBZHFTK-UHFFFAOYSA-N
Synonym 4-bromo-2-trifluoromethoxy thiophenol, 4-bromo-2-trifluoromethoxy benzenethiol, 4-bromo-2-trifluoromethoxy benzene-1-thiol, benzenethiol,4-bromo-2-trifluoromethoxy, pubchem2840, 4-bromo-2-trifluoromethoxy-benzenethiol, 4-bromanyl-2-trifluoromethyloxy benzenethiol
PubChem CID 2736425
IUPAC Name 4-bromo-2-(trifluoromethoxy)benzenethiol
SMILES FC(F)(F)OC1=C(S)C=CC(Br)=C1

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Quantity 10 g
Formula Weight 273.07
Percent Purity 95%
Chemical Name or Material 4-Bromo-2-(trifluoromethoxy)benzene-1-thiol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Bromo-2-(trifluoromethoxy)benzene-1-thiol, ≥95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.