missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-chloro-2-fluoropyridine, 98%, Thermo Scientific Chemicals

Product Code. 15401529
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Useful intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1017793-21-3
Molecular Formula C5H2BrClFN
Molecular Weight (g/mol) 210.43
MDL Number MFCD11040607
InChI Key UMQBGWKWOLRROL-UHFFFAOYSA-N
Synonym 4-bromo-3-chloro-2-fluoro-pyridine, pubchem22991, abbypharma ap-30-7506
PubChem CID 56604170
IUPAC Name 4-bromo-3-chloro-2-fluoropyridine
SMILES C1=CN=C(C(=C1Br)Cl)F

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 210.43
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Bromo-3-chloro-2-fluoropyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.