missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-fluoroacetophenone, 96%, Thermo Scientific Chemicals

Product Code. 15418638
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 304445-49-6
Molecular Formula C8H6BrFO
Molecular Weight (g/mol) 217.037
MDL Number MFCD00466241
InChI Key VCTWSAITPPCBHI-UHFFFAOYSA-N
Synonym 1-4-bromo-3-fluorophenyl ethanone, 4'-bromo-3'-fluoroacetophenone, 3-fluoro-4-bromo-acetophenone, 1-4-bromo-3-fluorophenyl ethan-1-one, 4-bromo-3-fluoroacetophenone, 4-bromo-3-fluoro-acetophenone, ksc915o9p, 3'-fluoro-4'-bromo-acetophenone, 1-4-bromo-3-fluoro-phenyl-ethanone
PubChem CID 22831914
IUPAC Name 1-(4-bromo-3-fluorophenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1)Br)F

Specifications

Quantity 250 mg
Formula Weight 217.04
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 4-Bromo-3-fluoroacetophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'