missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-nitrotoluene, 98+%, Thermo Scientific Chemicals

Product Code. 11469572
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5326-34-1
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.03
MDL Number MFCD00024180
InChI Key UPBUTKQMDPHQAQ-UHFFFAOYSA-N
Synonym 4-bromo-3-nitrotoluene, 2-bromo-5-methylnitrobenzene, benzene, 1-bromo-4-methyl-2-nitro, bromonitrotoluene-4, 1-bromo-4-methyl-2-nitro-benzene, 3-nitro-4-bromotoluene, pubchem9585, acmc-209l5d, ksc490m7t, tpc-i121
PubChem CID 79224
IUPAC Name 1-bromo-4-methyl-2-nitrobenzene
SMILES CC1=CC=C(Br)C(=C1)[N+]([O-])=O

Specifications

Melting Point 29°C to 32°C
Density 1.578
Boiling Point 96°C (1 mmHg)
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 1945925
Sensitivity Light sensitive
Formula Weight 216.04
Percent Purity ≥98%
Chemical Name or Material 4-Bromo-3-nitrotoluene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'