missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-4'-iodobiphenyl, 98%, Thermo Scientific Chemicals

Product Code. 15471519
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105946-82-5
Molecular Formula C12H8BrI
Molecular Weight (g/mol) 359.004
MDL Number MFCD17676204
InChI Key GWOAJJWBCSUGHH-UHFFFAOYSA-N
Synonym 4-bromo-4'-iodobiphenyl, 4-bromo-4'-iodo-1,1'-biphenyl, 1,1'-biphenyl, 4-bromo-4'-iodo, 1-bromo-4-4-iodophenyl benzene, 4'-bromo-4-iodobiphenyl, 4-iodo-4'-bromobiphenyl, 4-bromo-4'-iodo-biphenyl, 4-bromo-4/'-iodobiphenyl, ksc164g2f
PubChem CID 10904452
IUPAC Name 1-bromo-4-(4-iodophenyl)benzene
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)I)Br

Specifications

Melting Point 175°C to 179°C (literature)
Quantity 100 g
UN Number UN3152
Beilstein 2518914
Sensitivity Light sensitive
Formula Weight 359
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Bromo-4'-iodobiphenyl

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'