missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromoacetophenone, 98%, Thermo Scientific Chemicals

22.01 € - 212.06 €

Chemical Identifiers

CAS 99-90-1
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.05
MDL Number MFCD00000105
InChI Key WYECURVXVYPVAT-UHFFFAOYSA-N
Synonym 4'-bromoacetophenone, 1-4-bromophenyl ethanone, p-bromoacetophenone, 4-bromoacetophenone, ethanone, 1-4-bromophenyl, 1-4-bromophenyl ethan-1-one, 1-acetyl-4-bromobenzene, methyl p-bromophenyl ketone, p-bromophenyl methyl ketone, acetophenone, 4'-bromo
PubChem CID 7466
IUPAC Name 1-(4-bromophenyl)ethanone
SMILES CC(=O)C1=CC=C(Br)C=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11444903
View Documents
Thermo Scientific Alfa Aesar
A12222.14
25 g
22.01 €
25g
Please to purchase this item. Need a web account? Register with us today!
11454903
View Documents
Thermo Scientific Alfa Aesar
A12222.22
100 g
58.09 €
100g
Please to purchase this item. Need a web account? Register with us today!
11464903
View Documents
Thermo Scientific Alfa Aesar
A12222.36
500 g
212.06 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Labeled 4’-Bromoacetophenone possess activity against positive phototaxis of Chlamydomonas cells. A fundamental starting material for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Labeled 4’-Bromoacetophenone possess activity against positive phototaxis of Chlamydomonas cells. A fundamental starting material for organic synthesis.

Solubility
Soluble in Chloroform. Insoluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent, bases.
Specifications

Chemical Identifiers

99-90-1
199.05
WYECURVXVYPVAT-UHFFFAOYSA-N
7466
CC(=O)C1=CC=C(Br)C=C1
C8H7BrO
MFCD00000105
4'-bromoacetophenone, 1-4-bromophenyl ethanone, p-bromoacetophenone, 4-bromoacetophenone, ethanone, 1-4-bromophenyl, 1-4-bromophenyl ethan-1-one, 1-acetyl-4-bromobenzene, methyl p-bromophenyl ketone, p-bromophenyl methyl ketone, acetophenone, 4'-bromo
1-(4-bromophenyl)ethanone

Specifications

99-90-1
255°C to 256°C
C8H7BrO
386015
4'-bromoacetophenone, 1-4-bromophenyl ethanone, p-bromoacetophenone, 4-bromoacetophenone, ethanone, 1-4-bromophenyl, 1-4-bromophenyl ethan-1-one, 1-acetyl-4-bromobenzene, methyl p-bromophenyl ketone, p-bromophenyl methyl ketone, acetophenone, 4'-bromo
WYECURVXVYPVAT-UHFFFAOYSA-N
1-(4-bromophenyl)ethanone
7466
98%
49°C to 53°C
>110°C (230°F)
MFCD00000105
14,1403
Soluble in Chloroform. Insoluble in water.
CC(=O)C1=CC=C(Br)C=C1
199.05
199.05
4'-Bromoacetophenone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 202-799-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.