Learn More
4-(Bromoacetyl)pyridine hydrobromide, 98%, Thermo Scientific Chemicals
56.08 € - 571.85 €
Chemical Identifiers
CAS | 5349-17-7 |
---|---|
Molecular Formula | C7H7Br2NO |
Molecular Weight (g/mol) | 280.947 |
MDL Number | MFCD02681893 |
InChI Key | RGALBQILADNMKA-UHFFFAOYSA-N |
Synonym | 4-bromoacetyl pyridine hydrobromide, 2-bromo-1-pyridin-4-yl ethanone hydrobromide, 2-bromo-1-4-pyridinyl-1-ethanone hydrobromide, 2-bromo-1-pyridin-4-yl ethan-1-one hydrobromide, ethanone, 2-bromo-1-4-pyridinyl-, hydrobromide, 4-2-bromoacetyl pyridine hydrobromide, 4-bromoacetylpyridine hydrobromide, 2-bromo-1-pyrdin-4-ylethan-1-one hydrobromide, 2-bromo-1-4-pyridinyl-1-ethanone hbr, 4-bromoacetyl pyridine hbr |
PubChem CID | 2776239 |
IUPAC Name | 2-bromo-1-pyridin-4-ylethanone;hydrobromide |
SMILES | C1=CN=CC=C1C(=O)CBr.Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15434997
|
Thermo Scientific Alfa Aesar
H32285.03 |
1 g |
56.08 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15444997
|
Thermo Scientific Alfa Aesar
H32285.06 |
5 g |
175.88 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15454997
|
Thermo Scientific Alfa Aesar
H32285.14 |
25 g |
571.85 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Design and synthesis of potent, selective, and orally bioavailable tetrasubstituted imidazole inhibitors of p38 mitogen-activated protein kinase 4-(Bromoacetyl)pyridine hydrobromide was used to prepare 2-Phenyl-4-(4-pyridyl)imidazole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDesign and synthesis of potent, selective, and orally bioavailable tetrasubstituted imidazole inhibitors of p38 mitogen-activated protein kinase 4-(Bromoacetyl)pyridine hydrobromide was used to prepare 2-Phenyl-4-(4-pyridyl)imidazole.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
Chemical Identifiers
5349-17-7 | |
280.947 | |
RGALBQILADNMKA-UHFFFAOYSA-N | |
2776239 | |
C1=CN=CC=C1C(=O)CBr.Br |
C7H7Br2NO | |
MFCD02681893 | |
4-bromoacetyl pyridine hydrobromide, 2-bromo-1-pyridin-4-yl ethanone hydrobromide, 2-bromo-1-4-pyridinyl-1-ethanone hydrobromide, 2-bromo-1-pyridin-4-yl ethan-1-one hydrobromide, ethanone, 2-bromo-1-4-pyridinyl-, hydrobromide, 4-2-bromoacetyl pyridine hydrobromide, 4-bromoacetylpyridine hydrobromide, 2-bromo-1-pyrdin-4-ylethan-1-one hydrobromide, 2-bromo-1-4-pyridinyl-1-ethanone hbr, 4-bromoacetyl pyridine hbr | |
2-bromo-1-pyridin-4-ylethanone;hydrobromide |
Specifications
5349-17-7 | |
Weak | |
MFCD02681893 | |
4-bromoacetyl pyridine hydrobromide, 2-bromo-1-pyridin-4-yl ethanone hydrobromide, 2-bromo-1-4-pyridinyl-1-ethanone hydrobromide, 2-bromo-1-pyridin-4-yl ethan-1-one hydrobromide, ethanone, 2-bromo-1-4-pyridinyl-, hydrobromide, 4-2-bromoacetyl pyridine hydrobromide, 4-bromoacetylpyridine hydrobromide, 2-bromo-1-pyrdin-4-ylethan-1-one hydrobromide, 2-bromo-1-4-pyridinyl-1-ethanone hbr, 4-bromoacetyl pyridine hbr | |
RGALBQILADNMKA-UHFFFAOYSA-N | |
2-bromo-1-pyridin-4-ylethanone;hydrobromide | |
2776239 | |
98% |
∽200°C (decomposition) | |
C7H7Br2NO | |
UN3261 | |
Soluble in water. | |
C1=CN=CC=C1C(=O)CBr.Br | |
280.947 | |
280.95 | |
4-(Bromoacetyl)pyridine hydrobromide |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.