missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzenediazonium tetrafluoroborate, 96%

Product Code. 11401697
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11401697

Brand: Thermo Scientific Alfa Aesar B25670.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 673-40-5 | C6H4BBrF4N2 | 270.82 g/mol

4-Bromobenzenediazonium tetrafluoroborate was used in preparation of surface modified Au(111) electrode, 1-(4-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodec-1-ene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzenediazonium tetrafluoroborate was used in preparation of surface modified Au(111) electrode, 1-(4′-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodec-1-ene.

Notes
Moisture sensitive. Store away from oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 673-40-5
Molecular Formula C6H4BBrF4N2
Molecular Weight (g/mol) 270.82
MDL Number MFCD00011894
InChI Key FXTCQUCYDJZGGU-UHFFFAOYSA-N
Synonym 4-bromobenzenediazonium tetrafluoroborate, acmc-209nyn, 4-bromobenzene-1-diazonium; tetrafluoroboranuide, boron +3 cation; 4-bromobenzenediazonium; tetrafluoride
PubChem CID 2734810
IUPAC Name 4-bromobenzenediazonium;tetrafluoroborate
SMILES F[B-](F)(F)F.BrC1=CC=C(C=C1)[N+]#N

Specifications

Melting Point ∼135°C (decomposition)
Quantity 25 g
UN Number UN1759
Sensitivity Light and moisture sensitive
Formula Weight 270.83
Percent Purity 96%
Chemical Name or Material 4-Bromobenzenediazonium tetrafluoroborate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.