missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobutyl acetate, 97%, Thermo Scientific Chemicals

Product Code. 11457527
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4753-59-7
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.056
MDL Number MFCD00000263
InChI Key UOABIRUEGSGTSA-UHFFFAOYSA-N
Synonym 4-bromobutylacetate, 4-bromo-n-butyl acetate, 1-butanol, 4-bromo-, acetate, acetic acid 4-bromobutyl ester, 1-butanol, 4-bromo-, 1-acetate, 4-bromo-1-acetoxybutane, 1-butanol, 4-bromo-,1-acetate, bromobutyl acetate, 4-acetoxy-butylbromide, 4-acetoxybutyl bromide
PubChem CID 78491
IUPAC Name 4-bromobutyl acetate
SMILES CC(=O)OCCCCBr

Specifications

Density 1.348
Boiling Point 96°C to 97°C (15 mmHg)
Flash Point 109°C (228°F)
Refractive Index 1.46
Quantity 50 g
Beilstein 1749695
Formula Weight 195.07
Percent Purity ≥98%
Chemical Name or Material 4-Bromobutyl acetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'