Learn More
4-Bromobutyryl chloride, 95%, Thermo Scientific Chemicals
44.62 € - 132.66 €
Chemical Identifiers
CAS | 927-58-2 |
---|---|
Molecular Formula | C4H6BrClO |
Molecular Weight (g/mol) | 185.46 |
MDL Number | MFCD00000753 |
InChI Key | LRTRXDSAJLSRTG-UHFFFAOYSA-N |
Synonym | 4-bromobutyryl chloride, butanoyl chloride, 4-bromo, g-bromobutyroyl chloride, butyryl chloride, 4-bromo, 4-bromobutyrylchloride, bromobutryl chloride, bromobutyryl chloride, acmc-20ammt, 4-bromobutanoylchloride, 4-bromobutyroylchloride |
PubChem CID | 70225 |
IUPAC Name | 4-bromobutanoyl chloride |
SMILES | C(CC(=O)Cl)CBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10582841
|
Thermo Scientific Acros
303440050 |
5 g | Glass bottle |
44.62 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10351184
|
Thermo Scientific Acros
303440250 |
25 g | Glass bottle |
132.66 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
927-58-2 | |
185.46 | |
LRTRXDSAJLSRTG-UHFFFAOYSA-N | |
70225 | |
C(CC(=O)Cl)CBr |
C4H6BrClO | |
MFCD00000753 | |
4-bromobutyryl chloride, butanoyl chloride, 4-bromo, g-bromobutyroyl chloride, butyryl chloride, 4-bromo, 4-bromobutyrylchloride, bromobutryl chloride, bromobutyryl chloride, acmc-20ammt, 4-bromobutanoylchloride, 4-bromobutyroylchloride | |
4-bromobutanoyl chloride |
Specifications
927-58-2 | |
100.0 | |
1.6020g/mL | |
91°C | |
94% min. (GC) | |
Br(CH2)3COCl | |
MFCD00000753 | |
4-bromobutyryl chloride, butanoyl chloride, 4-bromo, g-bromobutyroyl chloride, butyryl chloride, 4-bromo, 4-bromobutyrylchloride, bromobutryl chloride, bromobutyryl chloride, acmc-20ammt, 4-bromobutanoylchloride, 4-bromobutyroylchloride | |
C(CC(=O)Cl)CBr | |
185.46 | |
185.46 | |
Liquid |
94.0 | |
Colorless to Yellow or Pink | |
101°C (37.0 mmHg) | |
Authentic | |
C4H6BrClO | |
1.4910 to 1.494 | |
1.602 | |
LRTRXDSAJLSRTG-UHFFFAOYSA-N | |
4-bromobutanoyl chloride | |
70225 | |
95% | |
4-Bromobutyryl chloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 213-154-
RUO – Research Use Only