missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobutyryl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11490013
Click to view available options
Quantity:
5 g
25 g
100 g

4-Bromobutyryl chloride is used as a precursor for the synthesis of carolic acid by reacting with ethoxymagnesiomalonic ester. It is widely utilized as an intermediate in organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobutyryl chloride is used as a precursor for the synthesis of carolic acid by reacting with ethoxymagnesiomalonic ester. It is widely utilized as an intermediate in organic synthesis and pharmaceuticals.

Solubility
Miscible with organic solvents. Immiscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents. Remove all sources of ignition. Keep the container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 927-58-2
Molecular Formula C4H6BrClO
Molecular Weight (g/mol) 185.445
MDL Number MFCD00000753
InChI Key LRTRXDSAJLSRTG-UHFFFAOYSA-N
Synonym 4-bromobutyryl chloride, butanoyl chloride, 4-bromo, g-bromobutyroyl chloride, butyryl chloride, 4-bromo, 4-bromobutyrylchloride, bromobutryl chloride, bromobutyryl chloride, acmc-20ammt, 4-bromobutanoylchloride, 4-bromobutyroylchloride
PubChem CID 70225
IUPAC Name 4-bromobutanoyl chloride
SMILES C(CC(=O)Cl)CBr

Specifications

Density 1.6
Boiling Point 102°C to 103°C (40 mmHg)
Flash Point 91°C (195°F)
Linear Formula Br(CH2)3COCl
Refractive Index 1.492
Quantity 100 g
UN Number UN3265
Beilstein 1743026
Sensitivity Moisture sensitive
Solubility Information Miscible with organic solvents. Immiscible with water.
Formula Weight 185.45
Percent Purity 97%
Chemical Name or Material 4-Bromobutyryl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromobutyryl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 4-Bromobutyryl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.