Learn More
4-Bromofuran-2-carboxamide, 96%, Thermo Scientific Chemicals
86.70 € - 952.00 €
Chemical Identifiers
CAS | 957345-95-8 |
---|---|
Molecular Formula | C5H4BrNO2 |
Molecular Weight (g/mol) | 189.996 |
MDL Number | MFCD12755858 |
InChI Key | UDNSRLLRTQICFU-UHFFFAOYSA-N |
Synonym | 4-bromo-furan-2-carboxylic acid amide |
PubChem CID | 57589975 |
IUPAC Name | 4-bromofuran-2-carboxamide |
SMILES | C1=C(OC=C1Br)C(=O)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15449027
|
Thermo Scientific Alfa Aesar
H52355.MD |
250 mg | N/A | ||||||
15429027
|
Thermo Scientific Alfa Aesar
H52355.03 |
1 g |
241.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15439027
|
Thermo Scientific Alfa Aesar
H52355.06 |
5 g |
952.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Reactant in the synthesis of 6-Phenanthridinones. Bromofuran-2-carboxamide used in the investigation of supramolecular amide and thioamide synthons in hydrogen bonding patterns.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsReactant in the synthesis of 6-Phenanthridinones. Bromofuran-2-carboxamide used in the investigation of supramolecular amide and thioamide synthons in hydrogen bonding patterns.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
957345-95-8 | |
189.996 | |
UDNSRLLRTQICFU-UHFFFAOYSA-N | |
57589975 | |
C1=C(OC=C1Br)C(=O)N |
C5H4BrNO2 | |
MFCD12755858 | |
4-bromo-furan-2-carboxylic acid amide | |
4-bromofuran-2-carboxamide |
Specifications
957345-95-8 | |
MFCD12755858 | |
UDNSRLLRTQICFU-UHFFFAOYSA-N | |
4-bromofuran-2-carboxamide | |
57589975 | |
96% |
C5H4BrNO2 | |
4-bromo-furan-2-carboxylic acid amide | |
C1=C(OC=C1Br)C(=O)N | |
189.996 | |
189.99 | |
4-Bromofuran-2-carboxamide |
Sicurezza e movimentazione
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only