missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoisophthalic acid, 96%, Thermo Scientific Chemicals

Product Code. 11404534
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6939-93-1
Molecular Formula C8H5BrO4
Molecular Weight (g/mol) 245.028
MDL Number MFCD00002404
InChI Key MSQIEZXCNYUWHN-UHFFFAOYSA-N
Synonym 4-bromoisophthalic acid, 1,3-benzenedicarboxylic acid, 4-bromo, 4-bromoisophthalicacid, 4-bromo-1,3-benzenedicarboxylic acid, 2-bromobenzene-1,5-dicarboxylic acid, pubchem12479, chemwish ic03530, ksc354c4d, msqiezxcnyuwhn-uhfffaoysa
PubChem CID 81361
IUPAC Name 4-bromobenzene-1,3-dicarboxylic acid
SMILES C1=CC(=C(C=C1C(=O)O)C(=O)O)Br

Specifications

Melting Point 296°C to 302°C
Quantity 5 g
Beilstein 1959392
Formula Weight 245.03
Percent Purity 96%
Chemical Name or Material 4-Bromoisophthalic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'