missing translation for 'onlineSavingsMsg'
Learn More
4-Bromoisophthalic acid, 96%, Thermo Scientific Chemicals Product Code.: 11424534

4-Bromoisophthalic acid, 96%, Thermo Scientific Chemicals

Product Code. 11424534
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11424534

Brand: Thermo Scientific Alfa Aesar A13152.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6939-93-1
Molecular Formula C8H5BrO4
Molecular Weight (g/mol) 245.028
MDL Number MFCD00002404
InChI Key MSQIEZXCNYUWHN-UHFFFAOYSA-N
Synonym 4-bromoisophthalic acid, 1,3-benzenedicarboxylic acid, 4-bromo, 4-bromoisophthalicacid, 4-bromo-1,3-benzenedicarboxylic acid, 2-bromobenzene-1,5-dicarboxylic acid, pubchem12479, chemwish ic03530, ksc354c4d, msqiezxcnyuwhn-uhfffaoysaShow MoreShow Less
PubChem CID 81361
IUPAC Name 4-bromobenzene-1,3-dicarboxylic acid
SMILES C1=CC(=C(C=C1C(=O)O)C(=O)O)Br

Specifications

Melting Point 296°C to 302°C
Quantity 100 g
Beilstein 1959392
Formula Weight 245.03
Percent Purity 96%
Chemical Name or Material 4-Bromoisophthalic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10243527 Certificate TypeCertificate of Analysis Date22/09/2022 Catalog NumberA13152.14,A13152.22,A13152.06
Lot Number10132425 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13152.14,A13152.22,A13152.06
Lot Number10216103 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13152.14,A13152.22,A13152.06
Lot NumberD9579A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13152.14,A13152.22,A13152.06
Lot Number10100771 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13152.14,A13152.22,A13152.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Bromoisophthalic acid, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.