missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromomethyl)tetrahydropyran, 97%, Thermo Scientific™

Product Code. 10378022
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 125552-89-8
Molecular Formula C6H11BrO
Molecular Weight (g/mol) 179.057
InChI Key LMOOYAKLEOGKJR-UHFFFAOYSA-N
Synonym 4-bromomethyl tetrahydropyran, 4-bromomethyltetrahydropyran, 4-bromomethyl oxane, 4-bromomethyl tetrahydro-2h-pyran, 4-bromomethyl-tetrahydro-pyran, 4-bromomethyl tetrahydro-pyran, 4-bromomethyl-tetrahydro-2h-pyran, 4-bromomethyl-tetrahydropyran, 2h-pyran, 4-bromomethyl tetrahydro
PubChem CID 2773286
IUPAC Name 4-(bromomethyl)oxane
SMILES C1COCCC1CBr

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 179.06
Percent Purity 97%
Chemical Name or Material 4-(Bromomethyl)tetrahydropyran
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'