missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenol, 97%

Product Code. 10645531 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
This item is not returnable. View return policy

Product Code. 10645531

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-41-2 | C6H5BrO | 173.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-41-2
Molecular Formula C6H5BrO
Molecular Weight (g/mol) 173.01
MDL Number MFCD00002313
InChI Key GZFGOTFRPZRKDS-UHFFFAOYSA-N
Synonym p-bromophenol, phenol, 4-bromo, p-bromohydroxybenzene, 4-bromo-phenol, p-bromophenic acid, phenol, p-bromo, para-bromophenol, 4-bromo phenol, unii-lao4j0183i, ccris 632
PubChem CID 7808
IUPAC Name 4-bromophenol
SMILES C1=CC(=CC=C1O)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 64°C to 68°C
Color Brown-Pink
Boiling Point 235°C to 236°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4OH
Quantity 100 g
Beilstein 06, 198
Merck Index 15, 1439
Solubility Information Solubility in water: 25g/L (25°C). Other solubilities: freely soluble in glacial acetic acid,freely soluble in alcohol,chloroform and ether
Formula Weight 173.01
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 4-Bromophenol
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 4-Bromophenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.