missing translation for 'onlineSavingsMsg'
Learn More
4-Bromophenyl isocyanate, 99%, Thermo Scientific Chemicals Product Code.: 11343016

4-Bromophenyl isocyanate, 99%, Thermo Scientific Chemicals

Product Code. 11343016
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11343016

Brand: Thermo Scientific Alfa Aesar L10834.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-Bromophenyl isocyanate is used in the synthesis of mixed bisamide derivative and monoamide product.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromophenyl isocyanate is used in the synthesis of mixed bisamide derivative and monoamide product.

Solubility
Sparingly soluble in water (0.40 g/L at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2493-02-9
Molecular Formula C7H4BrNO
Molecular Weight (g/mol) 198.02
MDL Number MFCD00002022
InChI Key CZQIJQFTRGDODI-UHFFFAOYSA-N
Synonym 4-bromophenyl isocyanate, 4-bromophenylisocyanate, p-bromophenyl isocyanate, benzene, 1-bromo-4-isocyanato, p-bromophenylisocyanate, p-bromocarbanil, p-bromophenyl carbonimide, isocyanic acid 4-bromophenyl ester, unii-mgr77m31zw, mgr77m31zw
PubChem CID 75609
IUPAC Name 1-bromo-4-isocyanatobenzene
SMILES C1=CC(=CC=C1N=C=O)Br

Specifications

Melting Point 39°C to 42°C
Boiling Point 157°C to 158°C (14 mmHg)
Flash Point 109°C (228°F)
Odor Pungent
Quantity 25 g
UN Number UN2811
Beilstein 508203
Sensitivity Moisture sensitive
Merck Index 14,1430
Solubility Information Sparingly soluble in water (0.40g/L at 25°C).
Formula Weight 198.03
Percent Purity 99%
Chemical Name or Material 4-Bromophenyl isocyanate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249858 Certificate TypeCertificate of Analysis Date02/04/2024 Catalog NumberL10834.06,L10834.14
Lot Number10245261 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog NumberL10834.06,L10834.14
Lot Number10241372 Certificate TypeCertificate of Analysis Date22/06/2022 Catalog NumberL10834.06,L10834.14
Lot Number10216702 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL10834.06,L10834.14
Lot Number10137322 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL10834.06,L10834.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Bromophenyl isocyanate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.