missing translation for 'onlineSavingsMsg'
Learn More

(4-Bromophenyl)methanesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10113314
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 53531-69-4
Molecular Formula C7H6BrClO2S
Molecular Weight (g/mol) 269.537
MDL Number MFCD04973454
InChI Key ZWVWFWGJZPHCHF-UHFFFAOYSA-N
Synonym 4-bromobenzylsulfonyl chloride, 4-bromophenyl methanesulfonyl chloride, 4-bromobenzylsulphonyl chloride, benzenemethanesulfonyl chloride, 4-bromo, 4-bromophenyl methanesulphonyl chloride, 4-bromophenyl methyl chlorosulfone, 4-bromo-phenyl-methanesulfonyl chloride, 4-bromo-benzenemethanesulfonyl chloride, pubchem2155, acmc-209l8j
PubChem CID 598764
IUPAC Name (4-bromophenyl)methanesulfonyl chloride
SMILES C1=CC(=CC=C1CS(=O)(=O)Cl)Br

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 269.55
Percent Purity 95%
Chemical Name or Material (4-Bromophenyl)methanesulfonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.