missing translation for 'onlineSavingsMsg'
Learn More

(4-Bromophenylethynyl)trimethylsilane, 98%, Thermo Scientific Chemicals

Product Code. 15401838
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as the intermediate of Organic Light Emitting Diode (OLED).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as the intermediate of Organic Light Emitting Diode (OLED).

Notes
Store at room temperature. Avoid strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16116-78-2
Molecular Formula C11H13BrSi
Molecular Weight (g/mol) 253.21
MDL Number MFCD01863556
InChI Key RNMSGCJGNJYDNS-UHFFFAOYSA-N
Synonym 4-bromophenylethynyl trimethylsilane, 4-bromophenyl ethynyl trimethylsilane, 4-bromo-phenylethynyl-trimethyl-silane, 2-4-bromophenyl ethynyl trimethylsilane, acmc-1bvav, p-bromophenylethynyl trimethylsilane, 4-trimethylsilylethynyl bromo benzene, 4-bromo-phenylethynyl trimethyl silane
PubChem CID 4226980
IUPAC Name 2-(4-bromophenyl)ethynyl-trimethylsilane
SMILES C[Si](C)(C)C#CC1=CC=C(Br)C=C1

Specifications

Melting Point 61°C to 63°C
Color White to Yellow
Physical Form Powder
Quantity 1 g
Formula Weight 253.21
Percent Purity 98%
Chemical Name or Material (4-Bromophenylethynyl)trimethylsilane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'