missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenylhydrazine hydrochloride, 99%

Product Code. 10581411 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 10581411

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 622-88-8 | C6H7BrN2·HCl | 223.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-88-8
Molecular Formula C6H7BrN2·HCl
Molecular Weight (g/mol) 223.51
MDL Number MFCD00012941
InChI Key RGGOWBBBHWTTRE-UHFFFAOYSA-N
Synonym 4-bromophenylhydrazine hydrochloride, 4-bromophenyl hydrazine hydrochloride, 4-bromophenylhydrazine hcl, 4-bromo phenyl hydrazine hydrochloride, p-bromophenylhydrazine monohydrochloride, hydrazine, 4-bromophenyl-, monohydrochloride, 1-4-bromophenyl hydrazine hydrochloride, 4-bromophenylhydrazinehydrochloride, p-bromophenylhydrazine hydrochloride
PubChem CID 12157
IUPAC Name (4-bromophenyl)hydrazine;hydrochloride
SMILES C1=CC(=CC=C1NN)Br.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 210°C to 215°C
Color Beige-Brown to Gray
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula BrC6H4NHNH2·HCl
Quantity 50 g
Beilstein 15, 434
Solubility Information Solubility in water: soluble
Formula Weight 223.51
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromophenylhydrazine hydrochloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromophenylhydrazine hydrochloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.