missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenylzinc iodide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Product Code. 15405365
Click to view available options
Quantity:
50 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 148651-39-2
Density 1.045
Molecular Formula C6H4BrIZn
MDL Number MFCD01311441
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Synonym 4-bromophenylzinc iodide, 4-bromophenylzinc iodide solution, 4-bromophenyl iodo zinc, 4-bromophenylzinc iodide 0.5 m in tetrahydrofuran, 4-bromophenylzinc iodide solution, 0.5 m in thf, 4-bromophenylzinc iodide, 0.5m in thf, packaged under argon in resealable chemseal bottles
InChI Key PLIIAGJFCAKUCV-UHFFFAOYSA-M
SMILES C1=CC(=CC=[C-]1)Br.[Zn+]I
IUPAC Name bromobenzene;iodozinc(1+)
Molecular Weight (g/mol) 348.286
PubChem CID 4595200
Formula Weight 348.29
Chemical Name or Material 4-Bromophenylzinc iodide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.