missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoresorcinol, 98%, Thermo Scientific Chemicals

89.95 € - 323.61 €

Chemical Identifiers

CAS 6626-15-9
Molecular Formula C6H5BrO2
Molecular Weight (g/mol) 189.008
MDL Number MFCD00002272
InChI Key MPCCNXGZCOXPMG-UHFFFAOYSA-N
Synonym 4-bromoresorcinol, 1-bromo-2,4-dihydroxybenzene, 1,3-benzenediol, 4-bromo, 4-bromo-1,3-dihydroxybenzene, 4-bromo-benzene-1,3-diol, 4-bromo-resorcinol, resorcinol, 4-bromo, acmc-209nuf, 2,4-dihydroxybromobenzene
PubChem CID 81105
IUPAC Name 4-bromobenzene-1,3-diol
SMILES C1=CC(=C(C=C1O)O)Br
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15482689
View Documents
Thermo Scientific Alfa Aesar
H66087.06
5 g
89.95 €
5g
Please to purchase this item. Need a web account? Register with us today!
15492689
View Documents
Thermo Scientific Alfa Aesar
H66087.14
25 g
323.61 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is commonly used as an organic reagent and pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is commonly used as an organic reagent and pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

6626-15-9
189.008
MPCCNXGZCOXPMG-UHFFFAOYSA-N
81105
C1=CC(=C(C=C1O)O)Br
C6H5BrO2
MFCD00002272
4-bromoresorcinol, 1-bromo-2,4-dihydroxybenzene, 1,3-benzenediol, 4-bromo, 4-bromo-1,3-dihydroxybenzene, 4-bromo-benzene-1,3-diol, 4-bromo-resorcinol, resorcinol, 4-bromo, acmc-209nuf, 2,4-dihydroxybromobenzene
4-bromobenzene-1,3-diol

Specifications

6626-15-9
Pink
Odorless
MFCD00002272
4-bromoresorcinol, 1-bromo-2,4-dihydroxybenzene, 1,3-benzenediol, 4-bromo, 4-bromo-1,3-dihydroxybenzene, 4-bromo-benzene-1,3-diol, 4-bromo-resorcinol, resorcinol, 4-bromo, acmc-209nuf, 2,4-dihydroxybromobenzene
MPCCNXGZCOXPMG-UHFFFAOYSA-N
4-bromobenzene-1,3-diol
81105
98%
4-Bromoresorcinol
97°C to 100°C
150°C (12 mmHg)
C6H5BrO2
1861280
Slightly soluble in water.
C1=CC(=C(C=C1O)O)Br
189.008
189.01
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'einecsNumber' : 229-586-8

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.