missing translation for 'onlineSavingsMsg'
Learn More

4-Butylbenzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10294974
Click to view available options
Quantity:
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 54997-92-1
Molecular Formula C10H13ClO2S
Molecular Weight (g/mol) 232.72
MDL Number MFCD00173908
InChI Key OVFZELSNOHIDEF-UHFFFAOYSA-N
Synonym 4-butylbenzene-1-sulfonyl chloride, 4-n-butylbenzenesulfonyl chloride, 4-n-butyl benzenesulphonyl chloride, 4-butyl-benzenesulfonyl chloride, 4-n-butylphenylsulfonyl chloride, 4-n-butylbenzenesulphonyl chloride, benzenesulfonyl chloride, 4-butyl, 4-butylphenyl chlorosulfone, pubchem5484, 4-n-butylbenzenesulfonylchloride
PubChem CID 2737352
SMILES CCCCC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3(CH2)3C6H4SO2Cl
Quantity 25 g
IUPAC Name 4-butylbenzene-1-sulfonyl chloride
Formula Weight 232.73
Percent Purity 97%
Chemical Name or Material 4-Butylbenzene-1-sulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'