missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Butylbenzene-1-sulfonyl chloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Linear Formula | CH3(CH2)3C6H4SO2Cl |
Quantity | 10 g |
IUPAC Name | 4-butylbenzenesulfonyl chloride |
Formula Weight | 232.73 |
Percent Purity | 97% |
Chemical Name or Material | 4-Butylbenzene-1-sulfonyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'