missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4-Carboxybutyl)triphenylphosphonium bromide, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Phosphonium salt is a key intermediate for manufacture of some types of prostaglandins, which are subsequently used as active ingredients of both human and veterinary drugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phosphonium salt is a key intermediate for manufacture of some types of prostaglandins, which are subsequently used as active ingredients of both human and veterinary drugs.
Solubility
Soluble in ethanol, methanol and soluble water. Insoluble in toluene and hexane.
Notes
Hygroscopic. Store in dry place. Keep away from oxidizing agents.
Phosphonium salt is a key intermediate for manufacture of some types of prostaglandins, which are subsequently used as active ingredients of both human and veterinary drugs.
Solubility
Soluble in ethanol, methanol and soluble water. Insoluble in toluene and hexane.
Notes
Hygroscopic. Store in dry place. Keep away from oxidizing agents.
Specifications
Specifications
Melting Point | 205°C to 209°C |
Flash Point | 195°C (383°F) |
Linear Formula | HO2C(CH2)4P(C6H5)3Br |
Quantity | 100 g |
Beilstein | 3586477 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in ethanol,methanol and soluble water. Insoluble in toluene and hexane. |
Formula Weight | 443.33 |
Percent Purity | 98% |
Chemical Name or Material | (4-Carboxybutyl)triphenylphosphonium bromide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'