missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-1,8-naphthalic anhydride, 94%
Click to view available options
Quantity:
25 g
100 g
500 g
missing translation for 'unitSize'
100g
25g
500g
Description
4-Chloro-1,8-naphthalic anhydride is used in dyes and pigments. It is also used in agrochemical, pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Chloro-1,8-naphthalic anhydride is used in dyes and pigments. It is also used in agrochemical, pharmaceutical.
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Store at 4°C. Store away from moisture. Incompatible with oxidizing agents, water, Strong bases, Strong acids.
4-Chloro-1,8-naphthalic anhydride is used in dyes and pigments. It is also used in agrochemical, pharmaceutical.
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Store at 4°C. Store away from moisture. Incompatible with oxidizing agents, water, Strong bases, Strong acids.
Specifications
Specifications
| Melting Point | 208°C to 214°C |
| Quantity | 25 g |
| Beilstein | 178072 |
| Sensitivity | Moisture sensitive |
| Synonym | 4-chloro-1,8-naphthalic anhydride, 4-chloronaphthalic anhydride, 6-chlorobenzo de isochromene-1,3-dione, 4-chloronapthalic anhydride, 6-chloro-1h,3h-benzo de isochromene-1,3-dione, 4-chloronaphthalene-1,8-dicarboxylic anhydride, 1h,3h-naphtho 1,8-cd pyran-1,3-dione, 6-chloro, naphthalic anhydride, 4-chloro-7ci,8ci, 6-chloro-1h,3h-naphtho 1,8-cd pyran-1,3-dione |
| Solubility Information | Hydrolyzes in water. |
| SMILES | C1=CC2=C(C=CC3=C2C(=C1)C(=O)OC3=O)Cl |
| PubChem CID | 77672 |
| Formula Weight | 232.63 |
| Percent Purity | 94% |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction