missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-fluorophenol, 98%, Thermo Scientific Chemicals

Product Code. 11468666
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 348-60-7
Molecular Formula C6H4ClFO
Molecular Weight (g/mol) 146.55
MDL Number MFCD00042583
InChI Key XLHYAEBESNFTCA-UHFFFAOYSA-N
Synonym 3-fluoro-4-chlorophenol, 4-chloro-3-fluoro-phenol, phenol, 4-chloro-3-fluoro, pubchem1493, acmc-1ctvv, 3-fluoro-4-chloro phenol, 3-fluoro-4-chloro-phenol, 3-fluoro-4-chlorophenol;, 4-chlor-3-fluor-phenol;, 4-chloro-3-fluorophenol;
PubChem CID 2724523
IUPAC Name 4-chloro-3-fluorophenol
SMILES OC1=CC=C(Cl)C(F)=C1

Specifications

Melting Point 50°C to 54°C
Boiling Point 84°C (44 mmHg)
Flash Point >110°C (230°F)
Odor Phenol-like
Quantity 5 g
UN Number UN3261
Beilstein 3236440
Formula Weight 146.55
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-fluorophenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.