missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-3-fluoropyridine hydrochloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Chloro-3-fluoropyridine hydrochloride is employed for the deprotection of phenol ethers.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
4-Chloro-3-fluoropyridine hydrochloride is employed for the deprotection of phenol ethers.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Specifications
Specifications
Melting Point | ∼160°C (sublimation) |
Quantity | 1 g |
Solubility Information | Soluble in water. |
Formula Weight | 168 |
Percent Purity | 97% |
Chemical Name or Material | 4-Chloro-3-fluoropyridine hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4-Chloro-3-fluoropyridine hydrochloride, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction